Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Varenicline tartrate | 375815-87-5 | MFCD08460603 | 99.9% | 361.35 | C17H19N3O6 | 50 MG
SDP

Catalog No. 5000215813
Encompass_Preferred

Varenicline tartrate is the tartrate salt of varenicline, a nicotinic acetylcholine receptor partial agonist. Supplied as a high-purity research chemical for in vitro and analytical applications, it is intended for laboratory use only and not for human or clinical use.

  • High purity (≈99.9%).
  • CAS number 375815-87-5.
  • Molecular weight 361.35 g/mol.
  • Appearance: off-white to yellow solid.
  • Available in multiple pack sizes, including small milligram quantities and larger bulk options.
  • Recommended storage: sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
  • Biological activity: partial agonist of α4β2 nicotinic acetylcholine receptors (IC50 ≈ 250 nM).

Catalog No. 50-002-15813 Supplier Medchemexpress LLC Supplier No. HY1002150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Varenicline tartrate is the tartrate salt of varenicline, a nicotinic acetylcholine receptor partial agonist. Supplied as a high-purity research chemical for in vitro and analytical applications, it is intended for laboratory use only and not for human or clinical use.

  • High purity (≈99.9%).
  • CAS number 375815-87-5.
  • Molecular weight 361.35 g/mol.
  • Appearance: off-white to yellow solid.
  • Available in multiple pack sizes, including small milligram quantities and larger bulk options.
  • Recommended storage: sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
  • Biological activity: partial agonist of α4β2 nicotinic acetylcholine receptors (IC50 ≈ 250 nM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.