Learn More
Cayman Chemical Varenicline (tartrate), Purity: ≥98%, CAS: 375815-87-5, 10 mg
SDP
Catalog No. 502969399
Encompass_Preferred
An agonist of α7 and α4β2 nAChRs (EC50s = 18 and 2.3 µM, respectively)
Catalog No. 50-296-9399 Supplier Cayman Chemical Supplier No. 1503010 MG
An agonist of α7 and α4β2 nAChRs (EC50s = 18 and 2.3 µM, respectively)