Learn More
Cayman Chemical VarenIclIntartrate 50mg
SDP
Catalog No. 502971891
Encompass_Preferred
An agonist of α7 and α4β2 nAChRs (EC50s = 18 and 2.3 µM, respectively)
Catalog No. 50-297-1891 Supplier Cayman Chemical Supplier No. 1503050 MG
An agonist of α7 and α4β2 nAChRs (EC50s = 18 and 2.3 µM, respectively)