Learn More
Selleck Chemical LLC Varespladib (LY315920) 50mg 172732-68-2
SDP
Catalog No. 508430
Encompass_Preferred
Varespladib (LY315920) is a potent and selective human non-pancreatic secretory phospholipase A2 (hnsPLA) inhibitor with IC50 of 7 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-843-0 Supplier Selleck Chemical LLC Supplier No. S111050MG
Varespladib (LY315920) is a potent and selective human non-pancreatic secretory phospholipase A2 (hnsPLA) inhibitor with IC50 of 7 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.