Learn More
Medchemexpress LLC Varespladib methyl | 172733-08-3 | 99.9% | 394.42 | 100 MG
SDP
Catalog No. 5000386225
Encompass_Preferred
Varespladib methyl is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2)
  • Orally-administered methyl ester pro-drug
  • High-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations
  • White to off-white solid appearance
  • Intended for research use only
Catalog No. 50-003-86225 Supplier Medchemexpress LLC Supplier No. HY17448100MG
Varespladib methyl is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2)
  • Orally-administered methyl ester pro-drug
  • High-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations
  • White to off-white solid appearance
  • Intended for research use only