Learn More
Medchemexpress LLC Varespladib methyl | 172733-08-3 | 99.9% | 394.42 | 25 MG
SDP
Catalog No. 5000384285
Encompass_Preferred
Varespladib methyl is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920. This prodrug demonstrates high-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations against 28 medically important snake venoms from six continents.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2).
  • Orally-administered methyl ester pro-drug.
  • Effective at nanomolar and picomolar concentrations against various snake venoms.
  • Available in various quantities as solid and solution forms.
Catalog No. 50-003-84285 Supplier Medchemexpress LLC Supplier No. 25MGHY17448
Varespladib methyl is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920. This prodrug demonstrates high-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations against 28 medically important snake venoms from six continents.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2).
  • Orally-administered methyl ester pro-drug.
  • Effective at nanomolar and picomolar concentrations against various snake venoms.
  • Available in various quantities as solid and solution forms.