Learn More
Medchemexpress LLC Varespladib methyl | 172733-08-3 | 99.9% | 394.42 | 50 MG
SDP
Catalog No. 5000386609
Encompass_Preferred
Varespladib methyl is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920. It demonstrates high-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations against 28 medically important snake venoms.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2)
  • Orally-administered methyl ester pro-drug
  • Metabolized to LY315920
  • Shows high-level sPLA2 inhibition against snake venoms
Catalog No. 50-003-86609 Supplier Medchemexpress LLC Supplier No. HY1744850MG
Varespladib methyl is a selective inhibitor of group II secretory phospholipase A2 (PLA2). It is an orally-administered methyl ester pro-drug that is metabolized to LY315920. It demonstrates high-level secretory PLA2 (sPLA2) inhibition at nanomolar and picomolar concentrations against 28 medically important snake venoms.
  • Selective inhibitor of group II secretory phospholipase A2 (PLA2)
  • Orally-administered methyl ester pro-drug
  • Metabolized to LY315920
  • Shows high-level sPLA2 inhibition against snake venoms