Learn More
Medchemexpress LLC Varespladib methyl | 172733-08-3 | 99.9% | 394.42 g/mol | C22H22N2O5 | 10 MG
SDP
Catalog No. 5000298395
Encompass_Preferred
Varespladib methyl is a small-molecule research compound that selectively inhibits group II secretory phospholipase A2 (sPLA2). It is supplied as a high-purity solid for biochemical and pharmacological studies, and is commonly used in in vitro enzyme inhibition assays, mechanistic studies of inflammatory pathways, and preclinical research on venom neutralization.
  • Selective inhibitor of group II secretory phospholipase A2 (sPLA2).
  • High purity suitable for research use (reported 99.90%).
  • Molecular formula C22H22N2O5; molecular weight 394.42 g/mol.
  • Available in small vial sizes for laboratory workflows.
  • Suitable for in vitro assays and preclinical studies involving PLA2 pathways.
Catalog No. 50-002-98395 Supplier Medchemexpress LLC Supplier No. HY1744810MG
Varespladib methyl is a small-molecule research compound that selectively inhibits group II secretory phospholipase A2 (sPLA2). It is supplied as a high-purity solid for biochemical and pharmacological studies, and is commonly used in in vitro enzyme inhibition assays, mechanistic studies of inflammatory pathways, and preclinical research on venom neutralization.
  • Selective inhibitor of group II secretory phospholipase A2 (sPLA2).
  • High purity suitable for research use (reported 99.90%).
  • Molecular formula C22H22N2O5; molecular weight 394.42 g/mol.
  • Available in small vial sizes for laboratory workflows.
  • Suitable for in vitro assays and preclinical studies involving PLA2 pathways.