Learn More
Cole-Parmer Digi-Sense Variable-Voltage Output Controller; 5-100%, 120V/10A
Catalog No. 501961556 Shop All Cole-Parmer Products
Encompass_Preferred
This control is designed to regulate the input voltage through the selector knob on the unit. To operate plug a resistive load into the receptacle on the back of the unit, and adjust the control dial to the desired level. The higher the selected number on the dial, the higher the voltage and hotter the load will get. This unit comes with a lattice support bracket, which is ideal for mounting to maximize space.
Catalog No. 50-196-1556 Supplier Cole-Parmer Supplier No. 3622559
This control is designed to regulate the input voltage through the selector knob on the unit. To operate plug a resistive load into the receptacle on the back of the unit, and adjust the control dial to the desired level. The higher the selected number on the dial, the higher the voltage and hotter the load will get. This unit comes with a lattice support bracket, which is ideal for mounting to maximize space.