Learn More
Perkin Elmer US LLC Variable Angle Relative Specular Reflectance Accessory
Catalog No. 502104145
Encompass_Preferred
The Variable Angle Reflectance Sample Holder is ideal for relative specular reflectance studies that require data from more than one angle of incidence. This accessory is compatible with the LAMBDA 25/35/45/365/650/750/850/850+/900/950/1050/1050+.
Catalog No. 50-210-4145 Supplier Perkin Elmer US LLC Supplier No. B0137314
Add to Cart
Add to Cart
Request a Quote
The Variable Angle Reflectance Sample Holder is ideal for relative specular reflectance studies that require data from more than one angle of incidence. This accessory is compatible with the LAMBDA 25/35/45/365/650/750/850/850+/900/950/1050/1050+.