Learn More
Grainger Variable Frequency Drive 1 hp 460V AC
Catalog No. 502750698
Encompass_Preferred
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V AC drive with a power rating of 7.5HP. It features sensorless vector control for applications requiring higher torque at low speed. The drive has built-in EMC filters and is suitable for use with asynchronous motors. Key features include PID control, flying start, fire mode, and sleep/wake-up mode. The drive can communicate via Modbus, CANopen, Ethernet IP, and other protocols. It measures 10.6 x 7.9 x 11.8 inches and weighs 17.6 pounds. The drive requires a dedicated circuit breaker and line reactor for optimal performance. It is designed for industrial automation applications requiring variable speed control.
Catalog No. 50-275-0698 Supplier Grainger Supplier No. 14H087
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V AC drive with a power rating of 7.5HP. It features sensorless vector control for applications requiring higher torque at low speed. The drive has built-in EMC filters and is suitable for use with asynchronous motors. Key features include PID control, flying start, fire mode, and sleep/wake-up mode. The drive can communicate via Modbus, CANopen, Ethernet IP, and other protocols. It measures 10.6 x 7.9 x 11.8 inches and weighs 17.6 pounds. The drive requires a dedicated circuit breaker and line reactor for optimal performance. It is designed for industrial automation applications requiring variable speed control.