Learn More
Grainger Variable Frequency Drive 15 hp 460V AC
Catalog No. 502750699
Encompass_Preferred
The Grainger website features a wide selection of variable frequency drives and accessories. These drives allow users to control AC motor speed and torque by varying the motor's input frequency and voltage. Offerings include VFDs from major brands like Allen-Bradley, Siemens, and Yaskawa with power ratings from 1/4 HP up to 500 HP. They have drives designed for applications like HVAC, pumps, and fans. Accessories include filters, reactors, bypass modules, and more to complement the VFDs. With various enclosure types and communication protocols, there are many configuration options to choose from to meet application needs.
Catalog No. 50-275-0699 Supplier Grainger Supplier No. 14H093
The Grainger website features a wide selection of variable frequency drives and accessories. These drives allow users to control AC motor speed and torque by varying the motor's input frequency and voltage. Offerings include VFDs from major brands like Allen-Bradley, Siemens, and Yaskawa with power ratings from 1/4 HP up to 500 HP. They have drives designed for applications like HVAC, pumps, and fans. Accessories include filters, reactors, bypass modules, and more to complement the VFDs. With various enclosure types and communication protocols, there are many configuration options to choose from to meet application needs.