Learn More
Grainger Variable Frequency Drive 20 hp 460V AC
Catalog No. 502750816
Encompass_Preferred
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V AC drive with a power rating of 7.5HP. It features sensorless vector control for applications requiring higher torque at low speed. The drive has built-in EMC filters and is suitable for variable torque applications like fans, pumps, and conveyors. It can be mounted vertically or horizontally without derating. The drive has a wide ambient operating temperature range of 14°F to 113°F and includes basic speed control, PID, and sleep mode. It can communicate via Modbus RTU, Ethernet IP, and other industrial protocols. The drive dimensions are 10.43" x 7.64" x 11.02" and it weighs 17 lbs.
Catalog No. 50-275-0816 Supplier Grainger Supplier No. 14H094
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V AC drive with a power rating of 7.5HP. It features sensorless vector control for applications requiring higher torque at low speed. The drive has built-in EMC filters and is suitable for variable torque applications like fans, pumps, and conveyors. It can be mounted vertically or horizontally without derating. The drive has a wide ambient operating temperature range of 14°F to 113°F and includes basic speed control, PID, and sleep mode. It can communicate via Modbus RTU, Ethernet IP, and other industrial protocols. The drive dimensions are 10.43" x 7.64" x 11.02" and it weighs 17 lbs.