Learn More
Grainger Variable Frequency Drive 25 hp 460V AC
Catalog No. 502750700
Encompass_Preferred
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V 3-phase AC drive with a power rating of 5 HP. It has an operating voltage range of 380-480V, can output 0-460V, and has a maximum output frequency of 500 Hz. This VFD drive has built-in PID control, Modbus communication, and can be programmed through its keypad and display. It offers overload protection, short circuit protection, and stall prevention. The drive has a compact IP20 rated enclosure and includes removable terminal blocks for easy wiring. It can provide energy savings by allowing variable speed control of 3-phase AC motors.
Catalog No. 50-275-0700 Supplier Grainger Supplier No. 14H095
Add to Cart
Add to Cart
Request a Quote
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V 3-phase AC drive with a power rating of 5 HP. It has an operating voltage range of 380-480V, can output 0-460V, and has a maximum output frequency of 500 Hz. This VFD drive has built-in PID control, Modbus communication, and can be programmed through its keypad and display. It offers overload protection, short circuit protection, and stall prevention. The drive has a compact IP20 rated enclosure and includes removable terminal blocks for easy wiring. It can provide energy savings by allowing variable speed control of 3-phase AC motors.