Learn More
Grainger Variable Frequency Drive 40 hp 460V AC
Catalog No. 502750807
Encompass_Preferred
The Schneider Electric Altivar 320 VFD is a 460V AC drive rated at 7.5HP, featuring sensorless vector control for high torque at low speeds. It includes EMC filters and suits variable torque applications like fans, pumps, and conveyors. It can be mounted vertically or horizontally without derating. Operating temperature ranges from 14°F to 113°F, with an altitude rating up to 3300 feet. Features include PID control, S-curve acceleration/deceleration, and fire mode. Programmable via display keypad or SoMove software, it is UL, cUL, and CE certified. Model: ATV320U07N4C.
Catalog No. 50-275-0807 Supplier Grainger Supplier No. 14H097
Add to Cart
Add to Cart
Request a Quote
The Schneider Electric Altivar 320 VFD is a 460V AC drive rated at 7.5HP, featuring sensorless vector control for high torque at low speeds. It includes EMC filters and suits variable torque applications like fans, pumps, and conveyors. It can be mounted vertically or horizontally without derating. Operating temperature ranges from 14°F to 113°F, with an altitude rating up to 3300 feet. Features include PID control, S-curve acceleration/deceleration, and fire mode. Programmable via display keypad or SoMove software, it is UL, cUL, and CE certified. Model: ATV320U07N4C.