Learn More
Grainger Variable Frequency Drive 5 hp 460V AC
Catalog No. 502750832
Encompass_Preferred
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V 3-phase AC drive with a power rating of 5 HP. It has a wide operating voltage range from 380V to 480V and can output 0 to 460V at its load. This VFD drive has built-in PID control, modbus communication, and can be configured for sensorless vector or V/Hz control modes. It is designed for variable torque applications like fans, pumps, and conveyors. The drive has a compact bookstyle design with front access to all control terminals which allows easy installation and wiring. It can provide overload protection and has built-in DC link reactors to minimize harmonics. The drive has cULus and NOM certification.
Catalog No. 50-275-0832 Supplier Grainger Supplier No. 14H090
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V 3-phase AC drive with a power rating of 5 HP. It has a wide operating voltage range from 380V to 480V and can output 0 to 460V at its load. This VFD drive has built-in PID control, modbus communication, and can be configured for sensorless vector or V/Hz control modes. It is designed for variable torque applications like fans, pumps, and conveyors. The drive has a compact bookstyle design with front access to all control terminals which allows easy installation and wiring. It can provide overload protection and has built-in DC link reactors to minimize harmonics. The drive has cULus and NOM certification.