Learn More
Grainger Variable Frequency Drive 7 1/2hp 460V
Catalog No. 502750776
Encompass_Preferred
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V AC drive with a power rating of 5 HP. It features sensorless vector control, an LCD display, and built-in EMC filtering. The drive has a wide ambient temperature operating range from 14°F to 113°F and includes RS485 Modbus communication. Safety features include short circuit and ground fault protection. The drive dimensions are 10.6 x 7.9 x 11.4 inches and it weighs 17.6 lbs.
Catalog No. 50-275-0776 Supplier Grainger Supplier No. 14H091
The Schneider Electric Altivar 320 Variable Frequency Drive is a 460V AC drive with a power rating of 5 HP. It features sensorless vector control, an LCD display, and built-in EMC filtering. The drive has a wide ambient temperature operating range from 14°F to 113°F and includes RS485 Modbus communication. Safety features include short circuit and ground fault protection. The drive dimensions are 10.6 x 7.9 x 11.4 inches and it weighs 17.6 lbs.