Learn More
Research Products International Corp Variable Height Tube Rack, 6 Tube Capacity
SDP
Catalog No. 50998156
Encompass
Features a flexible grip retainer that securely holds a combination of tube sizes from 1.5 ml up to 15 ml. Racks can be adjusted vertically to achieve the optimum depth of tube submersion in a water bath or beaker. Remains stable even when loaded with tubes of different size and weight. Convenient handle facilitates lifting rack in and out of a water bath without splashing. Autoclavable.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-998-156 Supplier Research Products International Corp Supplier No. 247510
Features a flexible grip retainer that securely holds a combination of tube sizes from 1.5 ml up to 15 ml. Racks can be adjusted vertically to achieve the optimum depth of tube submersion in a water bath or beaker. Remains stable even when loaded with tubes of different size and weight. Convenient handle facilitates lifting rack in and out of a water bath without splashing. Autoclavable.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.