Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Variable Micropipets
SDPSCI_ED

Catalog No. S90243E Shop All EDVOTEK Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 5000 μL
2 to 20 μL
20 to 200 μL
5 to 50 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S90243E 10 to 100 μL
S90243B 0.5 to 10 μL
S90243D 5 to 50 μL
S90243A 0.1 to 2.5 μL
S90243C 2 to 20 μL
S90243K 1000 to 5000 μL
S90243F 20 to 200 μL
S90243H 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S90243E Supplier Edvotek™ Supplier No. 591
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Service wrench
  • Instructions

Automatic tip ejector makes operation fast and convenient

EDVOTEK™'s new line of Variable Micropipets are smartly designed with volumes spanning 0.1 to 5000 μl. The contoured grip, lightweight and low force design, and large volume display make them easy-to-use. Sample volumes are selected by twisting the top and sample delivery utilizes standard micropipet tips. Each micropipet is fully autoclavable and calibratable. Make your laboratory experience authentic with these durable micropipets backed by a lifetime warranty.

Specifications

Autoclavable Autoclavable
Color Gray
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Plastic
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Variable Micropipet
Volume (Metric) 10 to 100 μL
Finger Style Ergonomic
Includes Service wrench, Instructions
Warranty Lifetime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.