Learn More
Waters Corp Waters Corporation Flowcell, Variable Pathlength, 0.15 to 3.00 mm, 1000 psi
Catalog No. 50792479 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This variable pathlength flowcell is designed for use in various laboratory applications. It offers flexibility with an adjustable pathlength and can withstand high pressures.
  • Features a variable pathlength design
  • Provides a pathlength range of 0.15 to 3.00 mm
  • Offers a volume range of 0.06 to 1.20 μL
  • Withstands a maximum pressure of 1000 psi
Catalog No. 50-792-479 Supplier Waters Corp Supplier No. 700000923
This variable pathlength flowcell is designed for use in various laboratory applications. It offers flexibility with an adjustable pathlength and can withstand high pressures.
  • Features a variable pathlength design
  • Provides a pathlength range of 0.15 to 3.00 mm
  • Offers a volume range of 0.06 to 1.20 μL
  • Withstands a maximum pressure of 1000 psi