Learn More
USA Scientific Inc Variable Speed 3D Rocker
Catalog No. 502745089
Encompass
Variable Speed 3D Rocker, 12 x 12 work surface, 2 to 30 rpm, includes one flat and one dimpled mat
Catalog No. 50-274-5089 Supplier USA Scientific Inc Supplier No. 25233230
Variable Speed 3D Rocker, 12 x 12 work surface, 2 to 30 rpm, includes one flat and one dimpled mat