Learn More
Perkin Elmer US LLC Variable Temperature Cell - Spectrum One, NaCl, 110 V, 60Hz
Supplier:  Perkin Elmer US LLC L1272290
Encompass_Preferred
The Variable Temperature Cell is used for the transmission study of solid or liquid samples at temperatures ranging from -190-250°C for solids and -70-250°C for liquids. These cells are compatible with the Frontier and Spectrum 0ne/100/400/65/Two/3 spectrometers.
Catalog No. 50-211-3525
Add to Cart
Request a Quote