Learn More
Perkin Elmer US LLC Variable Temperature Cell Window - KBr, Qty. 2
Catalog No. 502113527
Encompass_Preferred
The Variable Temperature Cell Holder is used in transmission studies of liquid or solid samples over a broad range of temperatures. The cell consists of a vacuum jacket that contains a refrigerant dewar/cell holder assembly and a heating block that contains the appropriate cell and allows variable temperature over a range of -190° to 250°C.
Catalog No. 50-211-3527 Supplier Perkin Elmer US LLC Supplier No. L1272291
The Variable Temperature Cell Holder is used in transmission studies of liquid or solid samples over a broad range of temperatures. The cell consists of a vacuum jacket that contains a refrigerant dewar/cell holder assembly and a heating block that contains the appropriate cell and allows variable temperature over a range of -190° to 250°C.