Learn More
Labsciences VARIABLE VOLUME PIPETTES.
Catalog No. 50821764
Encompass_Preferred
Variable volume pipettes. Volume Range 1 to 5 ml. Ergonomic, lightweight, streamline design. Click stop volume adjustment. Autoclavable nozzle. Supplied with a calibration tool and calibration report. Boxed.
Catalog No. 50-821-764 Supplier Labsciences Supplier No. 5000VV
Variable volume pipettes. Volume Range 1 to 5 ml. Ergonomic, lightweight, streamline design. Click stop volume adjustment. Autoclavable nozzle. Supplied with a calibration tool and calibration report. Boxed.