Learn More
Labsciences VARIABLE VOLUME PIPETTES.
Supplier:  Labsciences 50VV
Encompass_Preferred
Variable volume pipettes. Volume Range 5 to 50uL. Ergonomic, lightweight, streamline design. Click stop volume adjustment. Autoclavable nozzle. Supplied with a calibration tool and calibration report. Boxed.
Catalog No. 50-821-760