Learn More
Cole-Parmer Cole-Parmer Vortex Mixer, 0 to 3400 rpm, 115 VAC
Supplier:  Cole-Parmer 0472601
Encompass_Preferred
Shaker features two modes for both "touch" and continuous operation. The standard "Cup" head supplied with the vortexer will accommodate a single test tube. The head is easily switched with optional heads (sold separately) that accommodate tubes of a variety of sizes as well as microplates. No tools required. It is lightweight and stable during operation due to the optimized counter balance. Safe for cold room or incubator use.
Catalog No. NC0111903