Learn More
Perkin Elmer US LLC Variable-Angle Specular Reflectance Accessory
Catalog No. 502117255
Encompass_Preferred
The Variable-Angle Specular Reflectance Accessory can be used to obtain a spectrum of any flat, reflecting surface on plate or sheet material. Measurements of reflectance are useful in quality control of optical materials and for obtaining information about the crystalline structure of a material. The Variable-Angle Specular Reflectance Accessory can be used to: Measure reflection spectra of dielectrics at optimum angle Measure reflection-absorption spectra of thin films Measure refractive index and thickness of thin films Measure refractive indices from reflectance spectra using the Kramers-Kronig transformation
Catalog No. 50-211-7255 Supplier Perkin Elmer US LLC Supplier No. 01860954
Add to Cart
Add to Cart
Request a Quote
The Variable-Angle Specular Reflectance Accessory can be used to obtain a spectrum of any flat, reflecting surface on plate or sheet material. Measurements of reflectance are useful in quality control of optical materials and for obtaining information about the crystalline structure of a material. The Variable-Angle Specular Reflectance Accessory can be used to: Measure reflection spectra of dielectrics at optimum angle Measure reflection-absorption spectra of thin films Measure refractive index and thickness of thin films Measure refractive indices from reflectance spectra using the Kramers-Kronig transformation