Learn More
Inorganic Ventures VARIAN TUNING SOLUTION 125mL
SDP
Catalog No. 50723854
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Analyte: In, Be, Co, Mg, Th, Ba, Ce, Pb, and Tl 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-854 Supplier Inorganic Ventures Supplier No. VARTSMS125ML
Matrix: Nitric Acid (HNO3), Analyte: In, Be, Co, Mg, Th, Ba, Ce, Pb, and Tl 10 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search