Learn More
Inorganic Ventures VARIAN INTERNAL STD 125mL
SDP
Catalog No. 50723853
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Instrument(s): Agilent/Varian, Analyte: In, 6Li, Tb, Bi, Sc, and Y 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-853 Supplier Inorganic Ventures Supplier No. VARIS1125ML
Matrix: Nitric Acid (HNO3), Instrument(s): Agilent/Varian, Analyte: In, 6Li, Tb, Bi, Sc, and Y 100 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search