Learn More
Precision Plus Vacuum Parts Inc Varian, Agilent SH 100, SH110, Tip Seal Replacement
Catalog No. 501539897
Encompass_Preferred
Varian, Agilent, SH 100, SH 110, tip seal kits include tip seal & o-rings
Catalog No. 50-153-9897 Supplier Precision Plus Vacuum Parts Inc Supplier No. SH0100TS
Varian, Agilent, SH 100, SH 110, tip seal kits include tip seal & o-rings