Learn More
Creative Biomart Influenza A virus H3N2 (Panama) Antigen
Supplier:  Creative Biomart VAC0272
Encompass_Preferred
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Panama/2007/99. The Influenza Virus was purified by Ultracentrifugation with 10-40 % sucrose gradient.
Catalog No. 50-208-5112