Learn More
Sino Biological Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E Antibody, Rabbit MAb 20 µg
Catalog No. 5000749823
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E (Catalog# 40907-V08H; Q9J3M8; Met1-Tyr538). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-49823 Supplier Sino Biological Supplier No. 40907R14420
This antibody was obtained from a rabbit immunized with purified, recombinant Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E (Catalog# 40907-V08H; Q9J3M8; Met1-Tyr538). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.