Learn More
Sino Biological Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E Antibody, Rabbit PAb, Antigen Affinity Purified
Catalog No. 5000083722
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E ( Catalog#40907-V08H; Q9J3M8; Met1-Tyr538). gE/glycoprotein E specific IgG was purified by Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E affinity chromatography.
Catalog No. 50-000-83722 Supplier Sino Biological Supplier No. 40907T6220
Produced in rabbits immunized with purified, recombinant Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E ( Catalog#40907-V08H; Q9J3M8; Met1-Tyr538). gE/glycoprotein E specific IgG was purified by Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E affinity chromatography.