Learn More
Sino Biological Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer Antibody, Rabbit PAb, Antigen Affinity Purified 100 µL
Catalog No. 5000786605
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer (Catalog#CT125-VNAS; Q775J3 & Q9J3N1). Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer specific IgG was purified by Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer affinity chromatography.
Catalog No. 50-007-86605 Supplier Sino Biological Supplier No. CT125T62100
Produced in rabbits immunized with purified, recombinant Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer (Catalog#CT125-VNAS; Q775J3 & Q9J3N1). Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer specific IgG was purified by Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer affinity chromatography.