Learn More
Sino Biological Varicella-zoster virus (VZV) (strain Oka vaccine) gH & gL Heterodimer Protein, Biotinylated (MALS-verified) 20
Catalog No. 5000698760
Encompass_Preferred
It migrates as approximately 99.2 and 17.8 kDa band in SDS-PAGE under reducing conditions. The molecular weight of this protein is around 120.0 kDa verified by SEC-MALS.
Catalog No. 50-006-98760 Supplier Sino Biological Supplier No. CT125VNASB20
It migrates as approximately 99.2 and 17.8 kDa band in SDS-PAGE under reducing conditions. The molecular weight of this protein is around 120.0 kDa verified by SEC-MALS.