Learn More
Sino Biological Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B Antibody, Rabbit PAb, Antigen Affinity Purified 100 µL
Catalog No. 5000749839
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B (Catalog#40958-VNAS; Q4JR05). Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B specific IgG was purified by Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B affinity chromatography.
Catalog No. 50-007-49839 Supplier Sino Biological Supplier No. 40958T62100
Produced in rabbits immunized with purified, recombinant Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B (Catalog#40958-VNAS; Q4JR05). Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B specific IgG was purified by Varicella-zoster virus (VZV) (strain Oka vaccine) Glycoprotein B affinity chromatography.