Learn More
OHAUS Variety Set Model H301
Catalog No. 01804360 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-360 Vortex Attachment
1 options
Catalog No. 01-804-360 Supplier OHAUS Supplier No. 83041419
Add to Cart
Add to Cart
The Multiple Sample Starter Set (Model H301) can be used with the Vortex-Genie 2 mixers and the Vortex-Genie Pulse.
  • It consists of a 6 in. (152 mm) Platform 146-6005-00.
  • Sixty Microtube Foam Insert, and Microplate Foam Insert are also present.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse