Learn More
Lovibond™ VARIO Calmagite Hardness Set
Catalog No. 01671072 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
100 Tests
Catalog No. No. of Tests
01-671-072 100 Tests
1 options
This item is not returnable. View return policy
Catalog No. 01-671-072 Supplier Lovibond™ Supplier No. 535850
Add to Cart
Add to Cart
This item is not returnable. View return policy
Reagent set for determining the concentration of calcium and magnesium hardness in a water sample. This colorimetric test contains 4 liquid reagents for performing up to 100 tests.
  • Analyze calcium and magnesium hardness
  • Determine results with ease using this colorimetric test
  • Supplies all four reagents needed to perform the analysis
  • Range: 0.05 to 4 mg/L CaCO3 or Mg as MgCO3
Specifications
Analytes Calcium, Magnesium
For Use With (Application) Water Hardness Analysis
Includes Alkali Solution, Indicator Solution, EDTA Solution, EGTA Solution
No. of Tests 100 Tests
Sample Size 10 mL
Test Method Calmagite