Description
- The Vario product line was formulated for use in Lovibond test kits and instruments but can also be used in instruments and test kits from other manufacturer
- Easy to Open - Our powder packs have a defined Easy Open Notch that makes it simple to tear open the package.
- Easy to Dispense and Pour - Our powder packs easily create a V to pour the powder into the sample cell.
Specifications
Specifications
| pH Range | 0 to 14 |
| No. of Tests | 100 Tests |
| Type | Free Chlorine DPD |