Learn More
Vario DPD Buffer Solution - Total Chlorine, Lovibond™
Catalog No. 09050164 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
473 mL
Catalog No. Volume (Metric)
09-050-164 473 mL
1 options
Catalog No. 09-050-164 Supplier Tintometer Group Supplier No. 540223
Add to Cart
Add to Cart
Used for online chlorine monitoring, along with DPD buffer solution and DPD indicator powder.
  • Compatible with other manufacturers' online chlorine analyzers
  • Total Chlorine set part number: 540210
Specifications
pH Range 0 to 14
Volume (Metric) 473 mL
Type Total Chlorine Buffer