Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ VARIO EDTA Solution

Catalog No. 01671070 Shop All Tintometer Group Products
Change view
Click to view available options
Sample Size:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size
01-671-070 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-671-070 Supplier Lovibond™ Supplier No. 531470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liquid reagent used as part of a reagent set to determine the concentration of calcium and magnesium hardness in a water sample.

The VARIO product line was formulated for use in Lovibond™ test kits and instruments but can also be used in instruments and test kits from other manufacturers.

  • Liquid reagent dissolves and mixes easily into sample
  • High quality at an affordable price

Specifications

Analytes Calcium, Magnesium
Concentration 1 M
Volume (Metric) 50 mL
For Use With (Application) Water Hardness Analysis
Sample Size 10 mL
Test Method Calmagite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.