Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ VARIO Free Chlorine Solution

Catalog No. 01549788 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-788 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-788 Supplier Lovibond™ Supplier No. 531820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The VARIO Free Chlorine Reagent Solution uses the indophenol method to detect Free Chlorine in a water sample without the concern of interfering substances such as manganese and other oxidizing agents.

  • Direct detection of Free Chlorine in chloraminated samples
  • Liquid reagent dissolves and mixes easily into sample
  • This reagent should be used in combination with the Lovibond™ VARIO Monochlor F Reagent.
  • The VARIO product line was formulated for use in Lovibond™ test kits and instruments but can also be used in instruments and test kits from other manufacturers.

Specifications

Analytes Free Chlorine
Format Liquid Reagent
For Use With (Equipment) Select Lovibond™ Colorimeters and Spectrophotometers
No. of Tests ∼50
No. per Pack 30 mL
Quantity 1 Ea.
Range 0.02 to 4.50 mg/L Cl2
Sample Size 10 mL
Sterility Non-sterile
Test Method Lovibond™ M64
Test Time Varies Based on Sample Temperature: 5 to 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.