Learn More
Vario Molybdenum HR Reagent 2, F10, Lovibond™
Catalog No. 09050131 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
09-050-131 10 mL
1 options
Catalog No. 09-050-131 Supplier Tintometer Group Supplier No. 530760
Add to Cart
Add to Cart
Can be used with Lovibond colorimeters and test kits for water quality analysis, as well as those from other manufacturers.
  • The Vario product line was formulated for use in Lovibond test kits and instruments but can also be used in instruments and test kits from other manufacturer
  • Easy to Open - Our powder packs have a defined Easy Open Notch that makes it simple to tear open the package.
  • Easy to Dispense and Pour - Our powder packs easily create a V to pour the powder into the sample cell.
Specifications
pH Range 0 to 14
Volume (Metric) 10 mL
No. of Tests 100 Tests
Type Molybdenum Reagent