Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ VARIO Monochlor F Reagent

Catalog No. 01549787 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
50 to 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
01-549-787 50 to 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-787 Supplier Lovibond™ Supplier No. 531810
Add to Cart
Add to Cart
This item is not returnable. View return policy

The VARIO Monochlor F Reagent can directly detect the presence of monochloramines in a water sample using the indophenol method.

  • Direct detection of monochloramines with indophenol method
  • Easy to Handle - Lovibond™ powder packs are 37% larger than other manufacturers.
  • Easy to Open - The defined ''Easy Open Notch'' makes it simple to tear open the package.
  • Easy to Dispense and Pour - powder packs easily create a ''V'' to pour the powder into the sample cell.
  • The VARIO product line was formulated for use in Lovibond™ test kits and instruments but can also be used in instruments and test kits from other manufacturers.

Specifications

Analytes Monochloramines
Format Powder Pack
For Use With (Application) Water Samples From a Chloraminated System
For Use With (Equipment) Select Lovibond™ Colorimeters and Spectrophotometers
No. of Tests 50 to 100
No. per Pack 100
Packaging Zip-Lock Bag
Range 0.02 to 4.5 mg/L NH2Cl as Cl2 or 0.02 to 4.50 mg/L Cl2
Sample Size 10 mL
Sterility Non-sterile
Test Method Lovibond™ M63 and Lovibond™ M64
Test Time Varies Based on Sample Temperature: 5 to 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.