Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ VARIO Monochloramine Set

Catalog No. 01549790 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-790 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-790 Supplier Lovibond™ Supplier No. 535800
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure Monochlormines and Free Ammonia with one test! The VARIO Monochloramine Reagent Set uses the indophenol method which reduces testing complexity and saves time.

  • This set provides both reagents (one liquid and one powder pack) to detect Free Ammonia and Monochloramine in a single test.
  • VARIO Powder Pack Reagents are:
    • Easy to Handle - Lovibond™ powder packs are 37% larger than other manufacturers.
    • Easy to Open - The defined ''Easy Open Notch'' makes it simple to tear open the package.
    • Easy to Dispense and Pour - powder packs easily create a ''V'' to pour the powder into the sample cell.
  • The VARIO product line was formulated for use in Lovibond™ test kits and instruments but can also be used in instruments and test kits from other manufacturers.

Specifications

Analytes Monochloramines and Free Ammonia
For Use With (Application) Water Samples From a Chloraminated System
For Use With (Equipment) Select Lovibond™ Colorimeters and Spectrophotometers
Includes 1 x VARIO Monochlor F RGT Powder Pk., 1 x VARIO Free Ammonia Reagent Solution
No. of Tests ∼50
Quantity 1 Ea.
Range 0.02 to 4.5 mg/L NH2Cl as Cl2
Sample Size 10 mL
Test Method Lovibond™ M83
Test Time Varies Based on Sample Temperature: 5 to 15 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.