Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ VARIO NITRI NT-2 F10 Powder Pack Reagent

Catalog No. 01549785 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-785 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-785 Supplier Lovibond™ Supplier No. 530280
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use the VARIO NITRI NT-2 F10 Powder Pack reagent method to detect levels of Nitrite from 2 to 250 mg/L. These unit-dose reagents are easy to transport from site-to-site and make testing easy.

  • Ideal for measuring nitrite levels in closed-loop cooling systems.
  • Easy to Handle - Lovibond™ powder packs are 37% larger than other manufacturers.
  • Easy to Open - The defined ''Easy Open Notch'' makes it simple to tear open the package.
  • Easy to Dispense and Pour - powder packs easily create a ''V'' to pour the powder into the sample cell.
  • Monitoring nitrite levels in closed-loop cooling systems is key to preventing pipework corrosion. Use this VARIO Powder Pack reagent method to detect levels of Nitrite from 2 to 250 mg/L. These unit-dose reagents are easy to transport from site-to-site and make testing easy.
  • The VARIO product line was formulated for use in Lovibond™ test kits and instruments but can also be used in instruments and test kits from other manufacturers.

Specifications

Analytes Nitrite
Format Powder Pack Reagent
For Use With (Application) Closed-Loop Cooling Water Systems
For Use With (Equipment) Select Lovibond™ Colorimeters and Spectrophotometers
Includes 1 x VARIO NITRI NT-2 F10, 100 Pk.
No. of Tests 100
No. per Pack 100
Packaging Zip-Lock Bag
Quantity 1 Ea.
Range 2 to 250 mg/L NO2-
Sample Size 10 mL
Sterility Non-sterile
Test Method Lovibond™ M273
Test Time <15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.