Learn More
THERMO SCI LSG 3P Thermo Scientific Varioskan LUX, Absorbance, Fluorescence, Luminescence Reader, 1 ea
Supplier:  THERMO SCI LSG 3P VLBL0TD0
Encompass_Preferred
The Thermo Scientific™ Varioskan™ LUX Multimode Microplate Reader is a versatile, modular instrument for bioscience and life science research. This configuration offers Absorbance, Fluorescence Intensity, Luminescence, and Time-Resolved Fluorescence (TRF) measurement capabilities, providing high performance and precision for microplate-based assays.
  • Supports Absorbance, Fluorescence intensity (FRET), Luminescence, and Time-resolved fluorescence (TRF/hTRF)
  • Dual optics system with monochromator and filters for flexibility and sensitivity
  • Integrated gas control for setting CO2 and O2 concentrations
  • Simultaneous dispensing and measurement for real-time signal monitoring
Catalog No. 50-190-3783
Add to Cart
Request a Quote
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov