Learn More
THERMO SCI LSG 3P Thermo Fisher Microplate Reader, Varioskan LUX, No Dispenser, 1 Each
Supplier:  THERMO SCI LSG 3P VL0000D0
Encompass_Preferred
The Varioskan LUX is a modular multi-technology microplate reader controlled by SkanIt Software, providing comprehensive analytical capabilities for laboratory research.
  • Equipped with absorbance and fluorescence intensity detection.
  • Optional for luminescence, AlphaScreen, and time-resolved fluorescence.
  • Supports end point, kinetic, and spectral measurements.
  • Compatible with 6- to 1536-well microplates.
  • Features an incubator for temperature control up to 45°C.
  • Includes orbital shaking mode.
  • Xenon flash lamp light source.
  • Advanced optical system with dual monochromators.
  • Monochromator bandwidths: 5 nm/12 nm excitation, 12 nm emission.
Catalog No. 50-190-3762
Add to Cart
Request a Quote