Learn More
Medchemexpress LLC HY-10530 5mg Medchemexpress, Varlitinib CAS:845272-21-1 Purity:>98%
SDP
Catalog No. 501968266
Encompass_Preferred
Medchemexpress, HY-10530 5mg Varlitinib CAS:845272-21-1 Varlitinib (ARRY-334543; ASLAN001) is a potent, reversible, small molecule pan-EGFR inhibitor with IC50s of 7, 2, 4 nM for HER1, HER2 and HER4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8266 Supplier Medchemexpress LLC Supplier No. HY105305MG
Medchemexpress, HY-10530 5mg Varlitinib CAS:845272-21-1 Varlitinib (ARRY-334543; ASLAN001) is a potent, reversible, small molecule pan-EGFR inhibitor with IC50s of 7, 2, 4 nM for HER1, HER2 and HER4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.