Learn More
MSC Osborn Varnish Brush, 1-1/2 in Width, 1/2 in Thickness, White Bristle
Catalog No. 502839439
Encompass
Osborn varnish brushes are designed for general-purpose applications. Featuring white bristles, these brushes are suitable for applying various varnishes effectively.
  • 1-1/2 in brush width
  • 1/2 in thickness
  • 2-1/2 in trim length
  • White bristle material
  • Manufactured in the United States
Catalog No. 50-283-9439 Supplier MSC Supplier No. 53514840
Osborn varnish brushes are designed for general-purpose applications. Featuring white bristles, these brushes are suitable for applying various varnishes effectively.
  • 1-1/2 in brush width
  • 1/2 in thickness
  • 2-1/2 in trim length
  • White bristle material
  • Manufactured in the United States